Assay Details
| Gene Symbol: PCSK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000439.4 | NP_000430.3 | 12 - 13 | 1963 | 89 | |
| NM_001177875.1 | NP_001171346.1 | 12 - 13 | 1621 | 89 | ||
| GenBank mRNA | AB208874.1 | - | 12 - 13 | 1639 | 89 | |
| AK303888.1 | - | 12 - 13 | 1621 | 89 | ||
| AK308575.1 | - | 6 - 7 | 869 | 89 | ||
| BC130295.1 | - | 12 - 13 | 1845 | 40146326 | 89 | |
| BC136486.1 | - | 12 - 13 | 1845 | 9020475 | 89 | |
| M90753.1 | - | 12 - 13 | 1933 | 89 | ||
| X64810.1 | - | 12 - 13 | 1913 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001614.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | TC05001614.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | ENST00000311106.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | ENST00000508626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022025.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179790.hg.1 | TC0500011513.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000311106.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000508626.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000509190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | ENST00000513085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_000439.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011513.hg.1 | NM_001177875.1 | Refseq |

PCSK1
Genomic Map