Assay Details
| Gene Symbol: RIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256820.1 | NP_001243749.1 | 3 - 4 | 295 | 84 | |
| NM_001256821.1 | NP_001243750.1 | 4 - 5 | 322 | 84 | ||
| NM_006912.5 | NP_008843.1 | 4 - 5 | 444 | 84 | ||
| GenBank mRNA | AF084462.1 | - | 4 - 5 | 355 | 84 | |
| AF493923.1 | - | 3 - 4 | 241 | 84 | ||
| AK298768.1 | - | 3 - 4 | 295 | 84 | ||
| AK314239.1 | - | 4 - 5 | 420 | 84 | ||
| BC104186.2 | - | 4 - 5 | 315 | 40030939 | 84 | |
| BC104187.2 | - | 4 - 5 | 315 | 40030940 | 84 | |
| BI916790.1 | - | 4 - 5 | 305 | 84 | ||
| BM546985.1 | - | 4 - 5 | 438 | 84 | ||
| BQ918446.1 | - | 4 - 5 | 322 | 84 | ||
| CR407639.1 | - | 3 - 4 | 241 | 84 | ||
| U71203.1 | - | 4 - 5 | 376 | 84 | ||
| U78165.1 | - | 4 - 5 | 376 | 84 | ||
| Y07566.1 | - | 4 - 5 | 386 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003324.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | TC01003324.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | ENST00000368322.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | ENST00000368323.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | ENST00000461050.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | ENST00000539040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01051687.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100220795.hg.1 | TC0100015975.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368322.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000368323.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000461050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000462687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000539040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | ENST00000609492.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256820.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_001256821.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015975.hg.1 | NM_006912.5 | Refseq |

RIT1
Genomic Map