Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016063.2 | NP_057147.2 | 1 - 2 | 288 | 60 | |
| GenBank mRNA | AF151888.1 | - | 1 - 2 | 147 | 60 | |
| AF529363.1 | - | 1 - 2 | 84 | 60 | ||
| BC001671.2 | - | 1 - 2 | 223 | 60 | ||
| BC003357.1 | - | 1 - 2 | 127 | 60 | ||
| BC066332.1 | - | 1 - 2 | 102 | 5294646 | 60 | |
| BE743302.1 | - | 1 - 2 | 201 | 60 | ||
| DB447428.1 | - | 1 - 2 | 293 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002078.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06026007.hg.1 | TC06002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026007.hg.1 | ENST00000318787.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026007.hg.1 | ENST00000368377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026007.hg.1 | ENST00000398153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026007.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026008.hg.1 | TC06002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026008.hg.1 | ENST00000368377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026008.hg.1 | ENST00000398153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026008.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026006.hg.1 | TC06002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026006.hg.1 | ENST00000318787.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026006.hg.1 | ENST00000368377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026006.hg.1 | ENST00000398153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026006.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026005.hg.1 | TC06002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026005.hg.1 | ENST00000318787.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026005.hg.1 | ENST00000368377.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026005.hg.1 | ENST00000398153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026005.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190233.hg.1 | TC0600013073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190234.hg.1 | TC0600013073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190240.hg.1 | TC0600013073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600190242.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190242.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190242.hg.1 | TC0600013073.hg.1 | Clariom D TC |

HDDC2
Genomic Map