Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC001671.2 | - | 2 - 3 | 342 | 78 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |

HDDC2
Genomic Map