Assay Details
| Gene Symbol: KLHDC8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173546.2 | NP_775817.1 | 3 - 4 | 779 | 59 | |
| XM_005264940.4 | XP_005264997.1 | 3 - 4 | 900 | 59 | ||
| XM_006713016.2 | XP_006713079.1 | 3 - 4 | 1274 | 59 | ||
| GenBank mRNA | AY129011.1 | - | 3 - 4 | 736 | 59 | |
| BC039083.1 | - | 3 - 4 | 729 | 5167173 | 59 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000288.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005919.hg.1 | TC03000288.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005919.hg.1 | ENST00000332780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005919.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005919.hg.1 | ENST00000538729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005919.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300152323.hg.1 | TC0300007367.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000332780.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000459846.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000462582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000471811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | ENST00000476495.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300007367.hg.1 | NM_173546.2 | Refseq |

KLHDC8B
Genomic Map