Assay Details
| Gene Symbol: ARHGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005435.3 | NP_005426.2 | 14 - 15 | 4753 | 72 | |
| GenBank mRNA | AK090570.1 | - | 13 - 14 | 2203 | 72 | |
| AK123596.1 | - | 13 - 14 | 2952 | 72 | ||
| AK160365.1 | - | 14 - 15 | 4809 | 72 | ||
| BC010046.2 | - | 14 - 15 | 1698 | 72 | ||
| BC011839.2 | - | 14 - 15 | 1698 | 72 | ||
| BC014555.2 | - | 14 - 15 | 1698 | 72 | ||
| BC136661.1 | - | 14 - 15 | 4667 | 9020651 | 72 | |
| BX649178.1 | - | 13 - 14 | 2638 | 72 | ||
| U02082.1 | - | 12 - 13 | 1494 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000957.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
myeloid dendritic cell chemotaxis
actin cytoskeleton organization
regulation of actin cytoskeleton organization
regulation of Rho protein signal transduction
intracellular signal transduction
regulation of GTPase activity
positive regulation of JUN kinase activity
positive regulation of GTPase activity
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of cytoskeleton organization
positive regulation of stress fiber assembly
regulation of ERK1 and ERK2 cascade
positive regulation of podosome assembly
positive regulation of protein import
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
GTP binding
lipid binding
myeloid dendritic cell chemotaxis
actin cytoskeleton organization
regulation of actin cytoskeleton organization
regulation of Rho protein signal transduction
intracellular signal transduction
regulation of GTPase activity
positive regulation of JUN kinase activity
positive regulation of GTPase activity
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of cytoskeleton organization
positive regulation of stress fiber assembly
regulation of ERK1 and ERK2 cascade
positive regulation of podosome assembly
positive regulation of protein import
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
GTP binding
lipid binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014650.hg.1 | TC07000957.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014650.hg.1 | ENST00000056217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014650.hg.1 | ENST00000344879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014650.hg.1 | ENST00000471847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014650.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014649.hg.1 | TC07000957.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014649.hg.1 | ENST00000056217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014649.hg.1 | ENST00000344879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014649.hg.1 | ENST00000471847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07014649.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | ENST00000498580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700009536.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167735.hg.1 | TC0700009536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167734.hg.1 | TC0700009536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000056217.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000471847.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | ENST00000474817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | NM_005435.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167733.hg.1 | TC0700009536.hg.1 | Clariom D TC |

ARHGEF5
Genomic Map