Assay Details
| Gene Symbol: TYW5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039693.2 | NP_001034782.1 | 6 - 7 | 845 | 132 | |
| NR_004862.1 | 6 - 7 | 826 | 132 | |||
| NR_109907.1 | 5 - 6 | 690 | 132 | |||
| XM_017003310.1 | XP_016858799.1 | 4 - 5 | 473 | 132 | ||
| GenBank mRNA | AK095272.1 | - | 5 - 6 | 454 | 132 | |
| AK298977.1 | - | 6 - 7 | 589 | 132 | ||
| BC063502.1 | - | 2 - 3 | 114 | 132 | ||
| BC132835.1 | - | 6 - 7 | 609 | 40146857 | 132 | |
| BC136837.1 | - | 6 - 7 | 609 | 9020828 | 132 | |
| BC144420.1 | - | 6 - 7 | 590 | 132 | ||
| BX647677.1 | - | 6 - 7 | 827 | 133 | ||
| CN261486.1 | - | 6 - 7 | 558 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

TYW5
Genomic Map