Assay Details
| Gene Symbol: KIF5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004522.2 | NP_004513.1 | 13 - 14 | 1760 | 87 | |
| NR_111932.1 | 6 - 7 | 782 | 87 | |||
| XM_011511157.2 | XP_011509459.1 | 6 - 7 | 841 | 87 | ||
| XM_017004062.1 | XP_016859551.1 | 13 - 14 | 1751 | 87 | ||
| GenBank mRNA | AB011103.1 | - | 13 - 14 | 1733 | 87 | |
| AB073396.1 | - | 1 - 2 | 64 | 87 | ||
| AF010146.1 | - | 3 - 4 | 304 | 87 | ||
| BC110287.1 | - | 6 - 7 | 712 | 5165262 | 87 | |
| BC127772.1 | - | 10 - 11 | 1084 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000927.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
organelle organization
microtubule-based movement
axon guidance
motor neuron axon guidance
protein localization
cytoskeleton-dependent intracellular transport
mRNA transport
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
apolipoprotein receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | TC02000927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | ENST00000397413.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | ENST00000414838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | ENST00000435030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | ENST00000464066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013550.hg.1 | NM_004522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167252.hg.1 | TC0200009616.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000435030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000450621.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000460377.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000464066.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | ENST00000482151.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009616.hg.1 | NR_111932.1 | Refseq |

KIF5C
Genomic Map