Assay Details
| Gene Symbol: LYPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194317.3 | NP_919298.1 | 1 - 2 | 210 | 56 | |
| GenBank mRNA | AK309248.1 | - | 1 - 2 | 62 | 56 | |
| AY358623.1 | - | 1 - 2 | 126 | 56 | ||
| DA246384.1 | - | 1 - 2 | 211 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000929.hg
Additional Information:
This assay is in the 5'UTR region of the AY358623.1,NM_194317.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013608.hg.1 | TC02000929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013608.hg.1 | ENST00000334166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013608.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013616.hg.1 | TC02000929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013616.hg.1 | ENST00000334166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013616.hg.1 | ENST00000409381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013616.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013616.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013607.hg.1 | TC02000929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013607.hg.1 | ENST00000334166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013607.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167326.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167333.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167325.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167325.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167325.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167325.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167325.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167332.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 154938 | - | - |

LYPD6
Genomic Map