Assay Details
| Gene Symbol: CDH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004932.3 | NP_004923.1 | 4 - 5 | 995 | 101 | |
| XM_011513921.2 | XP_011512223.1 | 4 - 5 | 814 | 101 | ||
| XM_017008910.1 | XP_016864399.1 | 3 - 4 | 765 | 101 | ||
| XM_017008911.1 | XP_016864400.1 | 4 - 5 | 952 | 101 | ||
| GenBank mRNA | AK291290.1 | - | 4 - 5 | 961 | 101 | |
| BC000019.2 | - | 4 - 5 | 900 | 3506873 | 101 | |
| D31784.1 | - | 3 - 4 | 755 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000132.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05001273.hg.1 | TC05000132.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001273.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001273.hg.1 | ENST00000514738.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001273.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000508172.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000504835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000506396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000508172.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007044.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000265071.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000508132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | ENST00000514738.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | NM_004932.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500147541.hg.1 | TC0500007044.hg.1 | Clariom D TC |

CDH6
Genomic Map