Assay Details
| Gene Symbol: KDM4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014663.2 | NP_055478.2 | 14 - 15 | 2411 | 99 | |
| XM_005271354.3 | XP_005271411.1 | 14 - 15 | 2448 | 99 | ||
| XM_005271355.3 | XP_005271412.1 | 14 - 15 | 2366 | 99 | ||
| XM_005271356.3 | XP_005271413.1 | 15 - 16 | 2487 | 99 | ||
| XM_011542467.2 | XP_011540769.2 | 5 - 6 | 1519 | 99 | ||
| XM_017002912.1 | XP_016858401.1 | 15 - 16 | 2556 | 99 | ||
| GenBank mRNA | AB014577.1 | - | 14 - 15 | 2370 | 99 | |
| BC002558.2 | - | 14 - 15 | 2379 | 3138875 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000549.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
negative regulation of autophagy
negative regulation of gene expression
cardiac muscle hypertrophy in response to stress
viral process
histone demethylation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
negative regulation of histone H3-K9 trimethylation
protein binding
zinc ion binding
ubiquitin protein ligase binding
histone demethylase activity
methylated histone binding
histone demethylase activity (H3-K36 specific)
transcription, DNA-templated
negative regulation of autophagy
negative regulation of gene expression
cardiac muscle hypertrophy in response to stress
viral process
histone demethylation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
negative regulation of histone H3-K9 trimethylation
protein binding
zinc ion binding
ubiquitin protein ligase binding
histone demethylase activity
methylated histone binding
histone demethylase activity (H3-K36 specific)
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008885.hg.1 | TC01000549.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008885.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008885.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100157151.hg.1 | TC0100008073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000372396.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000463151.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000472265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000481296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | ENST00000485249.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008073.hg.1 | NM_014663.2 | Refseq |

KDM4A
Genomic Map