Assay Details
| Gene Symbol: QSOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004128.2 | NP_001004128.1 | 3 - 4 | 485 | 104 | |
| NM_002826.4 | NP_002817.2 | 3 - 4 | 485 | 104 | ||
| GenBank mRNA | AB209280.1 | - | 3 - 4 | 429 | 104 | |
| AF361868.1 | - | 3 - 4 | 435 | 104 | ||
| AK290842.1 | - | 3 - 4 | 434 | 104 | ||
| AK290887.1 | - | 3 - 4 | 449 | 104 | ||
| AY358941.1 | - | 3 - 4 | 436 | 104 | ||
| BC017692.1 | - | 3 - 4 | 424 | 4447666 | 104 | |
| BC100023.1 | - | 3 - 4 | 429 | 30563992 | 104 | |
| L42379.1 | - | 3 - 4 | 418 | 104 | ||
| U97276.2 | - | 3 - 4 | 485 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001559.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024122.hg.1 | TC01001559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024122.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024122.hg.1 | ENST00000367602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024122.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024122.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000443059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | ENST00000443059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010798.hg.1 | NR_026900.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024123.hg.1 | TC01001559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024123.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024123.hg.1 | ENST00000367602.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024123.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024123.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000443059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | ENST00000443059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010798.hg.1 | NR_026900.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | ENST00000392029.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179344.hg.1 | TC0100010798.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | ENST00000367600.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | ENST00000367602.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | NM_001004128.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | NM_002826.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179345.hg.1 | TC0100010798.hg.1 | Clariom D TC |

QSOX1
Genomic Map