Assay Details
| Gene Symbol: ADCK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174922.4 | NP_777582.4 | 5 - 6 | 629 | 79 | |
| XM_006716527.2 | XP_006716590.1 | 5 - 6 | 619 | 79 | ||
| XM_011516907.2 | XP_011515209.1 | 5 - 6 | 1342 | 79 | ||
| XM_011516909.1 | XP_011515211.1 | 5 - 6 | 629 | 79 | ||
| XM_011516910.1 | XP_011515212.1 | 6 - 7 | 652 | 79 | ||
| XM_017013174.1 | XP_016868663.1 | 5 - 6 | 667 | 79 | ||
| GenBank mRNA | AK092773.1 | - | 5 - 6 | 592 | 79 | |
| BC101659.1 | - | 5 - 6 | 569 | 8069165 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000851.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011357.hg.1 | TC08000851.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011357.hg.1 | ENST00000308860.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011357.hg.1 | ENST00000526231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08011357.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000525136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000526833.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000532190.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | ENST00000534714.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800009254.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000308860.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000526231.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000529654.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | ENST00000533715.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | NM_174922.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800163408.hg.1 | TC0800009254.hg.1 | Clariom D TC |

ADCK5
Genomic Map