Assay Details
| Gene Symbol: SLC30A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006345.3 | NP_006336.3 | 14 - 15 | 1394 | 63 | |
| XM_017007654.1 | XP_016863143.1 | 14 - 15 | 1428 | 63 | ||
| GenBank mRNA | AF006621.2 | - | 14 - 15 | 1368 | 63 | |
| AK225470.1 | - | 14 - 15 | 1331 | 63 | ||
| AK299917.1 | - | 15 - 16 | 1488 | 63 | ||
| AK312838.1 | - | 14 - 15 | 1394 | 63 | ||
| AY319413.1 | - | 15 - 16 | 1273 | 63 | ||
| AY594282.1 | - | 14 - 15 | 1248 | 63 | ||
| BC000240.1 | - | 14 - 15 | 1356 | 63 | ||
| BC007732.2 | - | 14 - 15 | 1355 | 4109702 | 63 | |
| BC016949.1 | - | 14 - 15 | 1373 | 3850521 | 63 | |
| BC022981.1 | - | 10 - 11 | 785 | 63 | ||
| KU178526.1 | - | 14 - 15 | 1248 | 63 | ||
| KU178527.1 | - | 14 - 15 | 1313 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000259.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleotide-excision repair
transcription, DNA-templated
zinc II ion transport
positive regulation of transcription from RNA polymerase II promoter
cation transmembrane transport
chromatin binding
transcription factor activity, sequence-specific DNA binding
cation transmembrane transporter activity
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
nucleotide-excision repair
transcription, DNA-templated
zinc II ion transport
positive regulation of transcription from RNA polymerase II promoter
cation transmembrane transport
chromatin binding
transcription factor activity, sequence-specific DNA binding
cation transmembrane transporter activity
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003832.hg.1 | TC04000259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003832.hg.1 | ENST00000264451.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003832.hg.1 | ENST00000536881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04003832.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000509683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000510460.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007361.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000264451.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000505523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | ENST00000513699.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | NM_006345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400150373.hg.1 | TC0400007361.hg.1 | Clariom D TC |

SLC30A9
Genomic Map