Assay Details
| Gene Symbol: NEURL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285485.1 | NP_001272414.1 | 2 - 3 | 612 | 132 | |
| NR_104346.1 | 2 - 3 | 801 | 132 | |||
| XM_011512178.2 | XP_011510480.1 | 3 - 4 | 1100 | 132 | ||
| XM_011512179.2 | XP_011510481.1 | 3 - 4 | 924 | 132 | ||
| GenBank mRNA | BC012317.1 | - | 2 - 3 | 801 | 132 | |
| HY288910.1 | - | 1 - 2 | 191 | 132 | ||
More Information
Set Membership: |
|

NEURL3
Genomic Map