Assay Details
| Gene Symbol: ZDHHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008222.2 | NP_001008223.1 | 7 - 8 | 1240 | 70 | |
| NM_016032.3 | NP_057116.2 | 8 - 9 | 1220 | 70 | ||
| GenBank mRNA | AB209807.1 | - | 7 - 8 | 1159 | 70 | |
| AF151847.1 | - | 8 - 9 | 1108 | 70 | ||
| AK001112.1 | - | 8 - 9 | 1170 | 70 | ||
| AK001424.1 | - | 5 - 6 | 554 | 70 | ||
| AK001524.1 | - | 7 - 8 | 971 | 70 | ||
| AL161962.1 | - | 8 - 9 | 1097 | 70 | ||
| AY358558.1 | - | 8 - 9 | 1152 | 70 | ||
| AY952881.8 | - | 8 - 9 | 1192 | 70 | ||
| BC000035.2 | - | 7 - 8 | 1114 | 3506601 | 70 | |
| BC003128.1 | - | 8 - 9 | 1158 | 2957911 | 70 | |
| BC006200.2 | - | 8 - 9 | 1154 | 2964425 | 70 | |
| BC012826.2 | - | 8 - 9 | 1154 | 2964425 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001355.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018139.hg.1 | TC0X001355.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018139.hg.1 | ENST00000357166.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018139.hg.1 | ENST00000371064.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018139.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018139.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018138.hg.1 | TC0X001355.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018138.hg.1 | ENST00000357166.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018138.hg.1 | ENST00000371064.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018138.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018138.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173202.hg.1 | TC0X00010793.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173201.hg.1 | TC0X00010793.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000357166.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000371064.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000406492.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000433917.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | ENST00000491039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_001008222.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010793.hg.1 | NM_016032.3 | Refseq |

ZDHHC9
Genomic Map