| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012412.hg.1 |
ENST00000475092.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012412.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200012412.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02028801.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02028801.hg.1 |
TC02001792.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200190663.hg.1 |
ENST00000475092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190663.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190663.hg.1 |
ENST00000475092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200190663.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200190663.hg.1 |
TC0200012412.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200012412.hg.1 |
ENST00000475092.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200012412.hg.1 |
NM_001101330.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200012412.hg.1 |
NM_001101330.2 |
Refseq |