Assay Details
| Gene Symbol: MAEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017405.2 | NP_001017405.1 | 7 - 8 | 959 | 94 | |
| NM_001297430.1 | NP_001284359.1 | 6 - 7 | 755 | 94 | ||
| NM_001297432.1 | NP_001284361.1 | 7 - 8 | 1097 | 94 | ||
| NM_001297433.1 | NP_001284362.1 | 7 - 8 | 1033 | 94 | ||
| NM_005882.4 | NP_005873.2 | 6 - 7 | 836 | 94 | ||
| NR_123716.1 | 5 - 6 | 759 | 94 | |||
| XM_006713849.3 | XP_006713912.1 | 7 - 8 | 919 | 94 | ||
| XM_006713850.2 | XP_006713913.1 | 7 - 8 | 1092 | 94 | ||
| XM_011513376.1 | XP_011511678.1 | 8 - 9 | 1079 | 94 | ||
| XM_011513377.1 | XP_011511679.1 | 7 - 8 | 997 | 94 | ||
| XM_017007636.1 | XP_016863125.1 | 8 - 9 | 1079 | 94 | ||
| XM_017007637.1 | XP_016863126.1 | 7 - 8 | 996 | 94 | ||
| GenBank mRNA | AB208907.1 | - | 2 - 3 | 3659 | 94 | |
| AF084928.1 | - | 7 - 8 | 797 | 94 | ||
| AK001088.1 | - | 7 - 8 | 919 | 94 | ||
| AK022586.1 | - | 6 - 7 | 796 | 94 | ||
| AK091951.1 | - | 2 - 3 | 830 | 94 | ||
| AK094253.1 | - | 4 - 5 | 580 | 94 | ||
| AK127875.1 | - | 3 - 4 | 1187 | 94 | ||
| AK296174.1 | - | 6 - 7 | 94 | |||
| AK298944.1 | - | 7 - 8 | 1013 | 94 | ||
| AK300572.1 | - | 8 - 9 | 856 | 94 | ||
| AK301155.1 | - | 7 - 8 | 927 | 94 | ||
| AY189687.1 | - | 7 - 8 | 872 | 94 | ||
| AY236486.1 | - | 5 - 6 | 704 | 94 | ||
| BC001225.1 | - | 7 - 8 | 926 | 3533990 | 94 | |
| BC006470.1 | - | 7 - 8 | 882 | 94 | ||
| BT006957.1 | - | 7 - 8 | 863 | 94 | ||
More Information
Set Membership: |
|

MAEA
Genomic Map