Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321314.1 | NP_001308243.1 | 4 - 5 | 1109 | 71 | |
| XM_005246586.1 | XP_005246643.1 | 5 - 6 | 1142 | 71 | ||
| XM_017004171.1 | XP_016859660.1 | 5 - 6 | 1515 | 71 | ||
| XM_017004172.1 | XP_016859661.1 | 5 - 6 | 1287 | 71 | ||
| XM_017004173.1 | XP_016859662.1 | 6 - 7 | 1370 | 71 | ||
| XM_017004174.1 | XP_016859663.1 | 5 - 6 | 1186 | 71 | ||
| GenBank mRNA | AK313542.1 | - | 4 - 5 | 1207 | 71 | |
| U47007.1 | - | 4 - 5 | 1109 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NAB1
Genomic Map