Assay Details
| Gene Symbol: NLRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134444.4 | NP_604393.2 | 5 - 6 | 2606 | 61 | |
| XM_017026345.1 | XP_016881834.1 | 4 - 5 | 2249 | 61 | ||
| GenBank mRNA | AF442488.1 | - | 4 - 5 | 2241 | 61 | |
| AF479747.1 | - | 4 - 5 | 2253 | 61 | ||
| AK056688.1 | - | 3 - 4 | 2133 | 61 | ||
| AK315003.1 | - | 5 - 6 | 2606 | 61 | ||
| AY072792.1 | - | 4 - 5 | 2244 | 61 | ||
| BC050326.1 | - | 5 - 6 | 2280 | 4837670 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000913.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19012593.hg.1 | ENST00000301295.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012593.hg.1 | ENST00000346986.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012593.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012593.hg.1 | TC19000913.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012592.hg.1 | ENST00000301295.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012592.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012592.hg.1 | TC19000913.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | TC1900008934.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170133.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | TC1900008934.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900170132.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000301295.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000587891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | ENST00000589437.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008934.hg.1 | NM_134444.4 | Refseq |

NLRP4
Genomic Map