Assay Details
| Gene Symbol: ADAMTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014244.4 | NP_055059.2 | 3 - 4 | 791 | 133 | |
| NM_021599.3 | NP_067610.1 | 3 - 4 | 791 | 133 | ||
| XM_017010063.1 | XP_016865552.1 | 2 - 3 | 227 | 133 | ||
| GenBank mRNA | AJ003125.1 | - | 3 - 4 | 689 | 133 | |
| AK308213.1 | - | 3 - 4 | 794 | 133 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05002130.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05029697.hg.1 | TC05002130.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029697.hg.1 | ENST00000251582.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029697.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029697.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05029697.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000518335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000522937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | ENST00000523450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013039.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | ENST00000251582.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | ENST00000274609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | NM_014244.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | NM_021599.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500191285.hg.1 | TC0500013039.hg.1 | Clariom D TC |

ADAMTS2
Genomic Map