Assay Details
| Gene Symbol: STXBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007269.2 | NP_009200.2 | 1 - 2 | 119 | 83 | |
| GenBank mRNA | AF032922.1 | - | 1 - 2 | 101 | 83 | |
| AK223170.1 | - | 1 - 2 | 98 | 83 | ||
| AK290134.1 | - | 1 - 2 | 103 | 83 | ||
| AK291322.1 | - | 1 - 2 | 99 | 83 | ||
| AK312910.1 | - | 1 - 2 | 96 | 83 | ||
| BC038099.2 | - | 1 - 2 | 58 | 4826366 | 83 | |
| BC047764.1 | - | 1 - 2 | 94 | 5585614 | 83 | |
| CN409056.1 | - | 1 - 2 | 143 | 83 | ||
| D63506.1 | - | 1 - 2 | 95 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000934.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vesicle docking involved in exocytosis
brain development
glucose transport
protein to membrane docking
insulin secretion
response to insulin
neutrophil degranulation
negative regulation of calcium ion-dependent exocytosis
protein heterooligomerization
platelet aggregation
protein binding
syntaxin-1 binding
syntaxin binding
vesicle docking involved in exocytosis
brain development
glucose transport
protein to membrane docking
insulin secretion
response to insulin
neutrophil degranulation
negative regulation of calcium ion-dependent exocytosis
protein heterooligomerization
platelet aggregation
protein binding
syntaxin-1 binding
syntaxin binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01014668.hg.1 | TC01000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014668.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014668.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014669.hg.1 | TC01000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014669.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014669.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014670.hg.1 | TC01000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014670.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014670.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014667.hg.1 | TC01000934.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014667.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01014667.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166463.hg.1 | TC0100009331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000472099.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000485167.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000486601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | ENST00000495245.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009331.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166464.hg.1 | TC0100009331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166461.hg.1 | TC0100009331.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000370008.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000469338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | ENST00000483586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | NM_007269.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100166465.hg.1 | TC0100009331.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 144171 | - | - |

STXBP3
Genomic Map