Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027953.hg.1 |
TC03001534.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027953.hg.1 |
ENST00000295569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027953.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03027953.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300185754.hg.1 |
TC0300011446.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000295569.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
ENST00000495737.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_001005527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011446.hg.1 |
NM_182522.4 |
Refseq |