Assay Details
| Gene Symbol: IL1R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261419.1 | NP_001248348.1 | 5 - 6 | 906 | 80 | |
| NM_004633.3 | NP_004624.1 | 5 - 6 | 906 | 80 | ||
| NR_048564.1 | 5 - 6 | 893 | 80 | |||
| XM_006712734.3 | XP_006712797.1 | 5 - 6 | 930 | 80 | ||
| XM_006712736.3 | XP_006712799.1 | 5 - 6 | 967 | 80 | ||
| XM_011511801.2 | XP_011510103.1 | 5 - 6 | 3266 | 80 | ||
| XM_011511803.2 | XP_011510105.1 | 5 - 6 | 2845 | 80 | ||
| XM_011511804.2 | XP_011510106.1 | 5 - 6 | 959 | 80 | ||
| XM_011511805.2 | XP_011510107.1 | 5 - 6 | 1732 | 80 | ||
| XM_017004889.1 | XP_016860378.1 | 4 - 5 | 816 | 80 | ||
| GenBank mRNA | AK130412.1 | - | 5 - 6 | 790 | 80 | |
| BC039031.1 | - | 5 - 6 | 906 | 5723005 | 80 | |
| FJ200480.1 | - | 4 - 5 | 517 | 80 | ||
| U64094.1 | - | 4 - 5 | 738 | 80 | ||
| U74649.1 | - | 4 - 5 | 712 | 80 | ||
| X59770.1 | - | 4 - 5 | 738 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000619.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009796.hg.1 | TC02000619.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009796.hg.1 | ENST00000332549.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009796.hg.1 | ENST00000393414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009796.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009796.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160461.hg.1 | TC0200008663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000332549.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000393414.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000441002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000457817.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000464994.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000474085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000482658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000485335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | ENST00000493749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_001261419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NM_004633.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008663.hg.1 | NR_048564.1 | Refseq |

IL1R2
Genomic Map