Assay Details
| Gene Symbol: SNX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013321.3 | NP_037453.1 | 6 - 7 | 833 | 98 | |
| XM_011515329.2 | XP_011513631.1 | 6 - 7 | 946 | 98 | ||
| XM_011515330.1 | XP_011513632.1 | 6 - 7 | 835 | 98 | ||
| XM_017012083.1 | XP_016867572.1 | 6 - 7 | 903 | 98 | ||
| XM_017012084.1 | XP_016867573.1 | 6 - 7 | 945 | 98 | ||
| GenBank mRNA | AF121858.1 | - | 6 - 7 | 789 | 98 | |
| AK074544.1 | - | 6 - 7 | 782 | 98 | ||
| AK315670.1 | - | 6 - 7 | 803 | 98 | ||
| BC007785.2 | - | 6 - 7 | 762 | 98 | ||
| BC021565.1 | - | 6 - 7 | 822 | 4564502 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001096.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07016895.hg.1 | TC07001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016895.hg.1 | ENST00000222990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016895.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016894.hg.1 | TC07001096.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016894.hg.1 | ENST00000222990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07016894.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | ENST00000617724.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013502.hg.1 | NR_106895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199742.hg.1 | TC0700013502.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000419693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000435060.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000457286.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000480807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000494722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | ENST00000617724.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013502.hg.1 | NR_106895.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | ENST00000222990.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | ENST00000479689.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | NM_013321.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700199743.hg.1 | TC0700013502.hg.1 | Clariom D TC |

SNX8
Genomic Map