Assay Details
| Gene Symbol: BCL2L14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017019963.1 | XP_016875452.1 | 2 - 3 | 162 | 157 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019963.1 transcripts.
Set Membership: |
|

BCL2L14
Genomic Map