Assay Details
| Gene Symbol: ATMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK301781.1 | - | 2 - 3 | 418 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK301781.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |

ATMIN
Genomic Map