Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016063.2 | NP_057147.2 | 2 - 3 | 413 | 88 | |
| GenBank mRNA | AF529363.1 | - | 2 - 3 | 209 | 88 | |
| BC003357.1 | - | 2 - 3 | 252 | 88 | ||
| BC066332.1 | - | 2 - 3 | 227 | 5294646 | 88 | |
| BE743302.1 | - | 2 - 3 | 326 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002078.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026003.hg.1 | TC06002078.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026003.hg.1 | ENST00000398153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06026003.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600190231.hg.1 | TC0600013073.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000318787.13 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000398153.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608284.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608461.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000608532.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609477.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | ENST00000609574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013073.hg.1 | NM_016063.2 | Refseq |

HDDC2
Genomic Map