| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026002.hg.1 |
TC06002078.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026002.hg.1 |
ENST00000318787.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026002.hg.1 |
ENST00000368377.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026002.hg.1 |
ENST00000398153.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06026002.hg.1 |
NM_016063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000318787.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000398153.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000608461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000608532.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000609477.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
NM_016063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000318787.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000398153.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000608461.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000608532.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
ENST00000609477.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
NM_016063.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600190227.hg.1 |
TC0600013073.hg.1 |
Clariom D TC |