Assay Details
| Gene Symbol: NRXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272020.1 | NP_001258949.1 | 5 - 6 | 1937 | 125 | |
| XM_005268218.2 | XP_005268275.1 | 22 - 23 | 5130 | 125 | ||
| XM_017021796.1 | XP_016877285.1 | 20 - 21 | 5079 | 125 | ||
| XM_017021799.1 | XP_016877288.1 | 21 - 22 | 5040 | 125 | ||
| GenBank mRNA | AK056530.1 | - | 5 - 6 | 1937 | 125 | |
More Information
Set Membership: |
|

NRXN3
Genomic Map