Assay Details
| Gene Symbol: GCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003765.1 | XP_016859254.1 | 1 - 2 | 455 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017003765.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |

GCA
Genomic Map