Assay Details
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006712400.3 | XP_006712463.1 | 2 - 3 | 456 | 90 | |
XM_017003764.1 | XP_016859253.1 | 2 - 3 | 469 | 90 | ||
XM_017003765.1 | XP_016859254.1 | 2 - 3 | 555 | 90 | ||
XM_017003766.1 | XP_016859255.1 | 2 - 3 | 352 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000233612.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000414723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000429691.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000437150.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000446271.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000453113.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000473240.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000479199.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000481161.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | ENST00000487445.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009829.hg.1 | NM_012198.4 | Refseq |
