Assay Details
| Gene Symbol: DAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199850.1 | NP_001186779.1 | 1 - 2 | 85 | 118 | |
| NM_004632.3 | NP_004623.1 | 1 - 2 | 82 | 118 | ||
| XM_011509972.2 | XP_011508274.1 | 1 - 2 | 81 | 118 | ||
| XM_017002291.1 | XP_016857780.1 | 2 - 3 | 170 | 118 | ||
| XM_017002294.1 | XP_016857783.1 | 2 - 3 | 170 | 118 | ||
| GenBank mRNA | AK293117.1 | - | 1 - 2 | 83 | 118 | |
| AK302281.1 | - | 1 - 2 | 85 | 118 | ||
| BC107487.1 | - | 1 - 2 | 82 | 40032516 | 118 | |
| X83544.1 | - | 1 - 2 | 67 | 118 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK293117.1, AK302281.1, BC107487.1, NM_001199850.1, NM_004632.3, X83544.1, XM_011509972.2, XM_017002291.1, XM_017002294.1 transcripts.
Set Membership: |
|

DAP3
Genomic Map