Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307923.1 | NP_001294852.1 | 5 - 6 | 666 | 119 | |
| NM_012397.3 | NP_036529.1 | 5 - 6 | 639 | 119 | ||
| XM_011526029.2 | XP_011524331.1 | 5 - 6 | 590 | 119 | ||
| GenBank mRNA | AF169949.1 | - | 4 - 5 | 501 | 119 | |
| AJ001696.2 | - | 5 - 6 | 573 | 119 | ||
| AJ001697.2 | - | 5 - 6 | 541 | 119 | ||
| AK290323.1 | - | 5 - 6 | 613 | 119 | ||
| AK296797.1 | - | 5 - 6 | 623 | 119 | ||
| AK303979.1 | - | 5 - 6 | 530 | 119 | ||
| AK313486.1 | - | 5 - 6 | 611 | 119 | ||
| BC101821.1 | - | 5 - 6 | 528 | 8069327 | 119 | |
| BC143402.1 | - | 5 - 6 | 555 | 9051909 | 119 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000228.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18002631.hg.1 | ENST00000269489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002631.hg.1 | ENST00000344731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002631.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002631.hg.1 | TC18000228.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002634.hg.1 | ENST00000269489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002634.hg.1 | ENST00000344731.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002634.hg.1 | ENST00000539341.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002634.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18002634.hg.1 | TC18000228.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | TC1800007547.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151790.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000415733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000431153.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | ENST00000479842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800007547.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | NM_012397.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | TC1800007547.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000269489.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000344731.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | ENST00000438844.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | NM_001307923.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800151788.hg.1 | NM_012397.3 | Refseq |

SERPINB13
Genomic Map