Assay Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199816.1 | NP_001186745.1 | 2 - 3 | 424 | 109 | |
| GenBank mRNA | BC005356.1 | - | 2 - 3 | 412 | 4102029 | 109 |
More Information
Set Membership: |
|

GGCT
Genomic Map