Assay Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199815.1 | NP_001186744.1 | 3 - 4 | 568 | 105 | |
| GenBank mRNA | BE407390.1 | - | 3 - 4 | 497 | 105 | |
More Information
Set Membership: |
|

GGCT
Genomic Map