Assay Details
| Gene Symbol: DSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005260909.4 | XP_005260966.1 | 4 - 5 | 1262 | 153 | |
| GenBank mRNA | AK300024.1 | - | 4 - 5 | 528 | 153 | |
| AK307465.1 | - | 3 - 4 | 273 | 153 | ||
| AK316460.1 | - | 4 - 5 | 444 | 153 | ||
More Information
Set Membership: |
|

DSCR3
Genomic Map