Assay Details
| Gene Symbol: DSCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005260911.4 | XP_005260968.1 | 2 - 3 | 968 | 144 | |
| GenBank mRNA | AK303624.1 | - | 2 - 3 | 399 | 144 | |
More Information
Set Membership: |
|

DSCR3
Genomic Map