Assay Details
| Gene Symbol: CLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF015601.1 | - | 3 - 4 | 319 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF015601.1 transcripts.
Set Membership: |
|

CLN3
Genomic Map