Assay Details
| Gene Symbol: PAPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002581.4 | NP_002572.2 | 17 - 18 | 4724 | 74 | |
| XM_006717129.2 | XP_006717192.1 | 15 - 16 | 2798 | 74 | ||
| XM_017014784.1 | XP_016870273.1 | 16 - 17 | 4650 | 74 | ||
| GenBank mRNA | AK300129.1 | - | 11 - 12 | 1655 | 74 | |
| BC078657.1 | - | 17 - 18 | 4354 | 30340336 | 74 | |
| X68280.1 | - | 17 - 18 | 4115 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006739.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006739.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006739.hg.1 | ENST00000534838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006739.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157165.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157165.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157165.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157165.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157165.hg.1 | TC0900008545.hg.1 | Clariom D TC |

PAPPA
Genomic Map