Assay Details
| Gene Symbol: PAPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002581.4 | NP_002572.2 | 20 - 21 | 5059 | 84 | |
| XM_006717129.2 | XP_006717192.1 | 18 - 19 | 3133 | 84 | ||
| XM_017014784.1 | XP_016870273.1 | 19 - 20 | 4985 | 84 | ||
| GenBank mRNA | AK300129.1 | - | 14 - 15 | 1990 | 84 | |
| BC078657.1 | - | 20 - 21 | 4689 | 30340336 | 84 | |
| X68280.1 | - | 20 - 21 | 4450 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09006742.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006742.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006742.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006742.hg.1 | ENST00000534838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006742.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006743.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006743.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006743.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006743.hg.1 | ENST00000534838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006743.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157168.hg.1 | TC0900008545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157169.hg.1 | TC0900008545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |

PAPPA
Genomic Map