Assay Details
| Gene Symbol: PAPPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002581.4 | NP_002572.2 | 7 - 8 | 3115 | 110 | |
| XM_006717129.2 | XP_006717192.1 | 5 - 6 | 1189 | 110 | ||
| XM_017014784.1 | XP_016870273.1 | 7 - 8 | 3155 | 110 | ||
| GenBank mRNA | AK300128.1 | - | 5 - 6 | 1342 | 110 | |
| BC078657.1 | - | 7 - 8 | 2745 | 30340336 | 110 | |
| X68280.1 | - | 7 - 8 | 2506 | 110 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006724.hg.1 | TC09000593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006724.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006724.hg.1 | ENST00000443904.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006724.hg.1 | ENST00000534838.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006724.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157153.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157153.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157153.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157153.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157153.hg.1 | TC0900008545.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000328252.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000460463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | ENST00000483254.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008545.hg.1 | NM_002581.4 | Refseq |

PAPPA
Genomic Map