Assay Details
| Gene Symbol: DNAH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277115.1 | NP_001264044.1 | 20 - 21 | 3883 | 79 | |
| GenBank mRNA | AJ320497.1 | - | 20 - 21 | 3883 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

DNAH11
Genomic Map