Assay Details
| Gene Symbol: DNAH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277115.1 | NP_001264044.1 | 35 - 36 | 6073 | 109 | |
| GenBank mRNA | AJ320497.1 | - | 35 - 36 | 6094 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

DNAH11
Genomic Map