Assay Details
| Gene Symbol: HERC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003922.3 | NP_003913.3 | 28 - 29 | 5393 | 103 | |
| XM_017022699.1 | XP_016878188.1 | 29 - 30 | 5499 | 103 | ||
| XM_017022700.1 | XP_016878189.1 | 29 - 30 | 5499 | 103 | ||
| XM_017022701.1 | XP_016878190.1 | 29 - 30 | 5499 | 103 | ||
| XM_017022702.1 | XP_016878191.1 | 28 - 29 | 5433 | 103 | ||
| XM_017022703.1 | XP_016878192.1 | 29 - 30 | 5499 | 103 | ||
| XM_017022704.1 | XP_016878193.1 | 29 - 30 | 5499 | 103 | ||
| XM_017022705.1 | XP_016878194.1 | 28 - 29 | 5406 | 103 | ||
| XM_017022706.1 | XP_016878195.1 | 29 - 30 | 5502 | 103 | ||
| GenBank mRNA | AK226162.1 | - | 15 - 16 | 2456 | 103 | |
| U50078.1 | - | 28 - 29 | 5341 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001544.hg.1
Set Membership: |
|

HERC1
Genomic Map