Assay Details
| Gene Symbol: HERC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003922.3 | NP_003913.3 | 34 - 35 | 6374 | 69 | |
| XM_017022699.1 | XP_016878188.1 | 35 - 36 | 6480 | 69 | ||
| XM_017022700.1 | XP_016878189.1 | 35 - 36 | 6480 | 69 | ||
| XM_017022701.1 | XP_016878190.1 | 35 - 36 | 6480 | 69 | ||
| XM_017022702.1 | XP_016878191.1 | 34 - 35 | 6414 | 69 | ||
| XM_017022703.1 | XP_016878192.1 | 35 - 36 | 6480 | 69 | ||
| XM_017022704.1 | XP_016878193.1 | 35 - 36 | 6480 | 69 | ||
| XM_017022705.1 | XP_016878194.1 | 34 - 35 | 6387 | 69 | ||
| XM_017022706.1 | XP_016878195.1 | 35 - 36 | 6483 | 69 | ||
| GenBank mRNA | AK226162.1 | - | 21 - 22 | 3437 | 69 | |
| U50078.1 | - | 34 - 35 | 6322 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001544.hg.1
Set Membership: |
|

HERC1
Genomic Map