Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014588.hg.1 |
ENST00000443617.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014588.hg.1 |
NM_003922.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014588.hg.1 |
TC15001544.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500171750.hg.1 |
ENST00000443617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171750.hg.1 |
NM_003922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171750.hg.1 |
ENST00000443617.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171750.hg.1 |
NM_003922.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171750.hg.1 |
TC1500009720.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007535.hg.1 |
ENST00000559303.2 |
Ensembl |