Assay Details
| Gene Symbol: ADGRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001908.1 | XP_016857397.1 | 3 - 4 | 1351 | 80 | |
| XM_017001909.1 | XP_016857398.1 | 3 - 4 | 1352 | 80 | ||
| XM_017001910.1 | XP_016857399.1 | 3 - 4 | 1350 | 80 | ||
| XM_017001911.1 | XP_016857400.1 | 3 - 4 | 1351 | 80 | ||
| GenBank mRNA | AK294999.1 | - | 2 - 3 | 355 | 80 | |
| AK296502.1 | - | 1 - 2 | 94 | 80 | ||
| AK307805.1 | - | 3 - 4 | 1120 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296502.1 transcripts.
Set Membership: |
|

ADGRB2
Genomic Map