Assay Details
| Gene Symbol: LBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194442.2 | NP_919424.1 | 1 - 2 | 153 | 76 | |
| GenBank mRNA | BE243907.1 | - | 1 - 2 | 224 | 76 | |
| DA447510.1 | - | 1 - 2 | 153 | 76 | ||
| DB091880.1 | - | 1 - 2 | 149 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003887.hg
Additional Information:
This assay is in the 5'UTR region of the NM_194442.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01059227.hg.1 | TC01003887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059227.hg.1 | ENST00000338179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059227.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059228.hg.1 | TC01003887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059228.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059223.hg.1 | TC01003887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059223.hg.1 | ENST00000272163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059223.hg.1 | ENST00000338179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059223.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059223.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232330.hg.1 | TC0100017488.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232335.hg.1 | TC0100017488.hg.1 | Clariom D TC |

LBR
Genomic Map