Assay Details
| Gene Symbol: LBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002296.3 | NP_002287.2 | 6 - 7 | 1007 | 88 | |
| NM_194442.2 | NP_919424.1 | 6 - 7 | 997 | 88 | ||
| XM_005273125.2 | XP_005273182.1 | 6 - 7 | 963 | 88 | ||
| XM_011544185.2 | XP_011542487.1 | 6 - 7 | 1409 | 88 | ||
| GenBank mRNA | AB209514.1 | - | 6 - 7 | 914 | 88 | |
| AK222834.1 | - | 6 - 7 | 894 | 88 | ||
| AK303589.1 | - | 6 - 7 | 913 | 88 | ||
| BC020079.1 | - | 6 - 7 | 929 | 3925138 | 88 | |
| L25931.1 | - | 6 - 7 | 908 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003887.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01059213.hg.1 | TC01003887.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059213.hg.1 | ENST00000272163.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059213.hg.1 | ENST00000338179.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059213.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01059213.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000421383.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000424022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000425080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000441022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000487054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | ENST00000488632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017488.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | ENST00000272163.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | ENST00000338179.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | NM_002296.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | NM_194442.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100232320.hg.1 | TC0100017488.hg.1 | Clariom D TC |

LBR
Genomic Map